ContactUsLinkComponent
Mouse Glutamate Receptor 4/GluA4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Glutamate Receptor 4/GluA4 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7277-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Glutamate Receptor 4/GluA4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Glutamate Receptor 4/GluA4 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7277-48
Supplier:  ANTIBODIES.COM