ContactUsLinkComponent
Human beta 1 Adrenergic Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human beta 1 Adrenergic Receptor in serum, plasma, tissue homogenates or other biological flu 1 * 96 Tests
Catalog # ANTIA82299-96
Supplier:  ANTIBODIES.COM
New Product
Human beta 1 Adrenergic Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human beta 1 Adrenergic Receptor in serum, plasma, tissue homogenates or other biological flu 1 * 96 Tests
Catalog # ANTIA82299-96
Supplier:  ANTIBODIES.COM