ContactUsLinkComponent
Human Casein Kinase 1 delta/CSNK1D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Casein Kinase 1 delta/CSNK1D in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7337-96
Supplier:  ANTIBODIES.COM
New Product
Human Casein Kinase 1 delta/CSNK1D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of human Casein Kinase 1 delta/CSNK1D in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7337-96
Supplier:  ANTIBODIES.COM