ContactUsLinkComponent
Porcine FGF21 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of porcine FGF21 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 96 Tests
Catalog # ANTIA5991-96
Supplier:  ANTIBODIES.COM
New Product
Porcine FGF21 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of porcine FGF21 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 96 Tests
Catalog # ANTIA5991-96
Supplier:  ANTIBODIES.COM