ContactUsLinkComponent
600102 Vacuum Controller VCpro 603 motor speed control and external sensor, 1100 - 1mbar, 115/230V, 50/60Hz with Schuko, UK & US plug leads 1 * 1 items
Catalog # ILMV600102
New Product
600102 Vacuum Controller VCpro 603 motor speed control and external sensor, 1100 - 1mbar, 115/230V, 50/60Hz with Schuko, UK & US plug leads 1 * 1 items
Catalog # ILMV600102