Order Entry
Belgium
Orders LinkContactUsLinkComponent
O411 98+%

O411 98+%

Bottle
Catalog # APOSOR21534-5G
Supplier:   Apollo Scientific
O411 98+%
Catalog # APOSOR21534-5G
Supplier:  Apollo Scientific
CAS Number:  175135-47-4
O411 98+%

Bottle

CAS Number:  175135-47-4
Price
2 910,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • MDL Number:MFCD00067778
  • Molecular Weight:253.30 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.