Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

O411 98+%

Bottle
Catalog # APOSOR21534-1G
Supplier:   Apollo Scientific
O411 98+%
Catalog # APOSOR21534-1G
Supplier:  Apollo Scientific
CAS Number:  175135-47-4
O411 98+%

Bottle

CAS Number:  175135-47-4
Price
833,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • MDL Number:MFCD00067778
  • Molecular Weight:253.30 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications