Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

O411 98+%

Bottle
Catalog # APOSOR21534-100MG
New Product
Supplier:   Apollo Scientific
O411 98+%
Catalog # APOSOR21534-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  175135-47-4
O411 98+%

Bottle

CAS Number:  175135-47-4
Price
181,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • MDL Number:MFCD00067778
  • Molecular Weight:253.30 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.