Order Entry
Belgium
Orders LinkContactUsLinkComponent
Selenium

Selenium

Bottle
Catalog # APOSIN3160-25G
Toxic
Health
Supplier:   Apollo Scientific
Selenium
Catalog # APOSIN3160-25G
Supplier:  Apollo Scientific
CAS Number:  
Selenium

Bottle

CAS Number:  
Price
43,50 €
Log in for availability
1 * 25 g
Product Details & Documents
  • Molecular Weight:78,95999 g/mol
  • UN:3283
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.