Order Entry
Belgium
Orders LinkContactUsLinkComponent
Valinomycin

Valinomycin

Bottle
Catalog # APOSBIV0893-1G
Toxique
Supplier:   Apollo Scientific
Valinomycin
Catalog # APOSBIV0893-1G
Supplier:  Apollo Scientific
CAS Number:  2001-95-8
Valinomycin

Bottle

CAS Number:  2001-95-8
Price
1 470,00 €
Connexion Pour la disponibilité
1 * 1 g
Product Details & Documents

Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions

  • Numéro CAS:2001-95-8
  • Formule:C₅₄H₉₀O₁₈N₆
  • Numéro MDL:MFCD00005114
  • Poids moléculaire:1111.34 g/mol
  • Point de fusion:187…190 °C
  • Température de stockage:Refrigerator
  • EINECS:217-896-6
  • UN:2811
  • ADR:6.1,I
Spécifications

Spécifications

  • Pack type:
    Bottle

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.