Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

DBCO-C6-NHS ester

Bottle
Catalog # APOSBIPG1892-1G
Irritant
New Product
Supplier:   Apollo Scientific
DBCO-C6-NHS ester
Catalog # APOSBIPG1892-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1384870-47-6
DBCO-C6-NHS ester

Bottle

CAS Number:  1384870-47-6
Price
1 380,00 €
Log in for availability
1 * 1 g
Product Details & Documents

DBCO-C6-NHS Ester is an amine-reactive compound, which can be used to modify an amine-containing molecule in organic media. This reagent has limited solubility in aqueous media. The extended 6-carbon atom spacer arm improves solubility in commonly used organic solvents including dichloromethane, chloroform, THF, and ethyl acetate and it also improves derivatization efficiency and stability of conjugates.

Synonyms:  DBCO-NHS ester 2, Dbco-c6-nhs ester
  • CAS Number:1384870-47-6
  • Formula:C₂₅H₂₂N₂O₅
  • MDL Number:MFCD22380763
  • Molecular Weight:430.453 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.