m-PEG24-NHS ester
Bottle
Catalog # APOSBIPG1631-250MG
New Product
Supplier: Apollo Scientific
m-PEG24-NHS ester
Catalog # APOSBIPG1631-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 756525-94-7
Jump to:
Product Details & Documents
- CAS Number:756525-94-7
- Formula:C₅₄H₁₀₃NO₂₈
- MDL Number:MFCD11041155
- Molecular Weight:1214.386 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



