Order Entry
Belgium
Orders LinkContactUsLinkComponent
m-PEG24-NHS ester

m-PEG24-NHS ester

Bottle
Catalog # APOSBIPG1631-1G
New Product
Supplier:   Apollo Scientific
m-PEG24-NHS ester
Catalog # APOSBIPG1631-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  756525-94-7
m-PEG24-NHS ester

Bottle

CAS Number:  756525-94-7
Price
1 900,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:756525-94-7
  • Formula:C₅₄H₁₀₃NO₂₈
  • MDL Number:MFCD11041155
  • Molecular Weight:1214.386 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.