Order Entry
Belgium
Orders LinkContactUsLinkComponent
m-PEG16-NHS ester

m-PEG16-NHS ester

Bottle
Catalog # APOSBIPG1630-50MG
New Product
Supplier:   Apollo Scientific
m-PEG16-NHS ester
Catalog # APOSBIPG1630-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
m-PEG16-NHS ester

Bottle

CAS Number:  
Price
521,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
  • Formula:C₃₈H₇₁NO₂₀
  • MDL Number:MFCD13184971
  • Molecular Weight:861.965 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.