Orders LinkContactUsLinkComponent
m-PEG16-NHS ester
Bottle
  APOSBIPG1630-250MG
New Product
m-PEG16-NHS ester
  APOSBIPG1630-250MG
New Product
 :  
m-PEG16-NHS ester
Bottle
 :  
1 500,00 €
1 * 250 mg
  •  :C₃₈H₇₁NO₂₀
  •  :MFCD13184971
  •  :861.965 g/mol
  •  :Ambient
  • Pack type:
    Bottle