m-PEG12-NHS ester
Bottle
Catalog # APOSBIPG1628-250MG
New Product
Supplier: Apollo Scientific
m-PEG12-NHS ester
Catalog # APOSBIPG1628-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 756525-94-7
Jump to:
Product Details & Documents
Synonyms: 2,5-Dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaoctatriacontan-38-oate, m-dPEG12-NHS-ester, mPEG12-NHS Ester, 4,7,10,13,16,19,22,25,28,31,34,37-Dodecaoxaoctatriacontanoic Acid N-Succinimidyl Ester, N-Succinimidyl 4,7,10,13,16,19,22,25,28,31,34,37-dodecaoxaoctatriacontanoate, Methyl-PEG12-NHS Ester
- CAS Number:756525-94-7
- Formula:C₃₀H₅₅NO₁₆
- MDL Number:MFCD13184970
- Molecular Weight:685.755 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



