Order Entry
Belgium
Orders LinkContactUsLinkComponent
m-PEG12-NHS ester

m-PEG12-NHS ester

Bottle
Catalog # APOSBIPG1628-10G
New Product
Supplier:   Apollo Scientific
m-PEG12-NHS ester
Catalog # APOSBIPG1628-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  756525-94-7
m-PEG12-NHS ester

Bottle

CAS Number:  756525-94-7
Price
3 400,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  2,5-Dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaoctatriacontan-38-oate, m-dPEG12-NHS-ester, mPEG12-NHS Ester, 4,7,10,13,16,19,22,25,28,31,34,37-Dodecaoxaoctatriacontanoic Acid N-Succinimidyl Ester, N-Succinimidyl 4,7,10,13,16,19,22,25,28,31,34,37-dodecaoxaoctatriacontanoate, Methyl-PEG12-NHS Ester
  • CAS Number:756525-94-7
  • Formula:C₃₀H₅₅NO₁₆
  • MDL Number:MFCD13184970
  • Molecular Weight:685.755 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.