Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

m-PEG3-NHS ester

Bottle
Catalog # APOSBIPG1621-250MG
New Product
Supplier:   Apollo Scientific
m-PEG3-NHS ester
Catalog # APOSBIPG1621-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
m-PEG3-NHS ester

Bottle

CAS Number:  
Price
163,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
  • Formula:C₁₂H₁₉NO₇
  • MDL Number:MFCD20228789
  • Molecular Weight:289.282 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications