Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mal-PEG6-NHS ester

Mal-PEG6-NHS ester

Bottle
Catalog # APOSBIPG1507-5G
New Product
Supplier:   Apollo Scientific
Mal-PEG6-NHS ester
Catalog # APOSBIPG1507-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mal-PEG6-NHS ester

Bottle

CAS Number:  
Price
3 420,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,5-dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12,15,18-hexaoxahenicosan-21-oate
  • Formula:C₂₃H₃₄N₂O₁₂
  • MDL Number:MFCD20926374
  • Molecular Weight:530.522 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.