Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mal-PEG6-NHS ester

Bottle
Catalog # APOSBIPG1507-1G
New Product
Supplier:   Apollo Scientific
Mal-PEG6-NHS ester
Catalog # APOSBIPG1507-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mal-PEG6-NHS ester

Bottle

CAS Number:  
Price
1 140,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2,5-dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12,15,18-hexaoxahenicosan-21-oate
  • Formula:C₂₃H₃₄N₂O₁₂
  • MDL Number:MFCD20926374
  • Molecular Weight:530.522 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications