Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mal-PEG3-NHS ester

Mal-PEG3-NHS ester

Bottle
Catalog # APOSBIPG1504-5G
New Product
Supplier:   Apollo Scientific
Mal-PEG3-NHS ester
Catalog # APOSBIPG1504-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mal-PEG3-NHS ester

Bottle

CAS Number:  
Price
2 170,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)ethoxy)propanoate
  • Formula:C₁₇H₂₂N₂O₉
  • MDL Number:MFCD26127782
  • Molecular Weight:398.365 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications