Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mal-PEG2-NHS ester

Mal-PEG2-NHS ester

Bottle
Catalog # APOSBIPG1503-250MG
New Product
Supplier:   Apollo Scientific
Mal-PEG2-NHS ester
Catalog # APOSBIPG1503-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mal-PEG2-NHS ester

Bottle

CAS Number:  
Price
322,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)propanoate
  • Formula:C₁₅H₁₈N₂O₈
  • MDL Number:MFCD24539465
  • Molecular Weight:354.312 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.