Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mal-PEG1-NHS ester

Bottle
Catalog # APOSBIPG1502-5G
New Product
Supplier:   Apollo Scientific
Mal-PEG1-NHS ester
Catalog # APOSBIPG1502-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mal-PEG1-NHS ester

Bottle

CAS Number:  
Price
3 570,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,5-Dioxopyrrolidin-1-yl 3-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)propanoate
  • Formula:C₁₃H₁₄N₂O₇
  • MDL Number:MFCD26127783
  • Molecular Weight:310.259 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications