Orders LinkContactUsLinkComponent
Mal-PEG1-NHS ester
Bottle
  APOSBIPG1502-250MG
New Product
Mal-PEG1-NHS ester
  APOSBIPG1502-250MG
New Product
 :  
Mal-PEG1-NHS ester
Bottle
 :  
396,00 €
1 * 250 mg
 :  2,5-Dioxopyrrolidin-1-yl 3-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)propanoate
  •  :C₁₃H₁₄N₂O₇
  •  :MFCD26127783
  •  :310.259 g/mol
  •  :Ambient
  • Pack type:
    Bottle