Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fmoc-PEG8-NHS ester

Fmoc-PEG8-NHS ester

Bottle
Catalog # APOSBIPG1421-5G
Supplier:   Apollo Scientific
Fmoc-PEG8-NHS ester
Catalog # APOSBIPG1421-5G
Supplier:  Apollo Scientific
CAS Number:  
Fmoc-PEG8-NHS ester

Bottle

CAS Number:  
Price
1 040,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • Formula:C₃₈H₅₂N₂O₁₄
  • MDL Number:MFCD21363318
  • Molecular Weight:760.824 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.