DNP-PEG4-NHS ester
Bottle
Catalog # APOSBIPG1388-250MG
New Product
Supplier: Apollo Scientific
DNP-PEG4-NHS ester
Catalog # APOSBIPG1388-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 858126-78-0
Jump to:
Product Details & Documents
Synonyms: 2,5-Dioxopyrrolidin-1-yl 1-((2,4-dinitrophenyl)amino)-3,6,9,12-tetraoxapentadecan-15-oate
- CAS Number:858126-78-0
- Formula:C₂₁H₂₈N₄O₁₂
- MDL Number:MFCD13184987
- Molecular Weight:528.467 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



