Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bis-PEG3-NHS ester

Bis-PEG3-NHS ester

Bottle
Catalog # APOSBIPG1264-5G
New Product
Supplier:   Apollo Scientific
Bis-PEG3-NHS ester
Catalog # APOSBIPG1264-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1314378-16-9
Bis-PEG3-NHS ester

Bottle

CAS Number:  1314378-16-9
Price
2 050,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Bis-dPEG®3-NHS ester
  • CAS Number:1314378-16-9
  • Formula:C₁₈H₂₄N₂O₁₁
  • Molecular Weight:444.39 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.