Orders LinkContactUsLinkComponent
Bis-PEG3-NHS ester
Bottle
  APOSBIPG1264-250MG
New Product
Bis-PEG3-NHS ester
  APOSBIPG1264-250MG
New Product
 :  1314378-16-9
Bis-PEG3-NHS ester
Bottle
 :  1314378-16-9
223,00 €
1 * 250 mg
 :  Bis-dPEG®3-NHS ester
  •  :1314378-16-9
  •  :C₁₈H₂₄N₂O₁₁
  •  :444.39 g/mol
  •  :Ambient
  • Pack type:
    Bottle