Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bis-PEG3-NHS ester

Bis-PEG3-NHS ester

Bottle
Catalog # APOSBIPG1264-1G
New Product
Supplier:   Apollo Scientific
Bis-PEG3-NHS ester
Catalog # APOSBIPG1264-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1314378-16-9
Bis-PEG3-NHS ester

Bottle

CAS Number:  1314378-16-9
Price
507,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Bis-dPEG®3-NHS ester
  • CAS Number:1314378-16-9
  • Formula:C₁₈H₂₄N₂O₁₁
  • Molecular Weight:444.39 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications