Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bis-PEG1-NHS ester

Bottle
Catalog # APOSBIPG1262-250MG
New Product
Supplier:   Apollo Scientific
Bis-PEG1-NHS ester
Catalog # APOSBIPG1262-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Bis-PEG1-NHS ester

Bottle

CAS Number:  
Price
225,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  Bis(2,5-dioxopyrrolidin-1-yl) 3,3'-oxydipropanoate
  • Formula:C₁₄H₁₆N₂O₉
  • MDL Number:MFCD20226397
  • Molecular Weight:356.285 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications