Order Entry
Belgium
Orders LinkContactUsLinkComponent
Biotin-PEG8-NHS ester

Biotin-PEG8-NHS ester

Bottle
Catalog # APOSBIPG1231-1G
Supplier:   Apollo Scientific
Biotin-PEG8-NHS ester
Catalog # APOSBIPG1231-1G
Supplier:  Apollo Scientific
CAS Number:  2143968-03-8
Biotin-PEG8-NHS ester

Bottle

CAS Number:  2143968-03-8
Price
1 750,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  N-Succinimidyl 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoate, 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 29-oxo-33-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13,16,19,22,25-octaoxa-28-azatritriacontanoate
  • CAS Number:2143968-03-8
  • Formula:C₃₃H₅₆N₄O₁₄S
  • MDL Number:MFCD27977502
  • Molecular Weight:764.89 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications