Biotin-PEG8-NHS ester
Bottle
Catalog # APOSBIPG1231-100MG
Supplier: Apollo Scientific
Biotin-PEG8-NHS ester
Catalog # APOSBIPG1231-100MG
Supplier: Apollo Scientific
CAS Number: 2143968-03-8
Jump to:
Product Details & Documents
Synonyms: N-Succinimidyl 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoate, 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 29-oxo-33-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13,16,19,22,25-octaoxa-28-azatritriacontanoate
- CAS Number:2143968-03-8
- Formula:C₃₃H₅₆N₄O₁₄S
- MDL Number:MFCD27977502
- Molecular Weight:764.89 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



