Order Entry
Belgium
Orders LinkContactUsLinkComponent
Biotin-PEG3-NHS ester

Biotin-PEG3-NHS ester

Bottle
Catalog # APOSBIPG1229-1G
New Product
Supplier:   Apollo Scientific
Biotin-PEG3-NHS ester
Catalog # APOSBIPG1229-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1253286-56-4
Biotin-PEG3-NHS ester

Bottle

CAS Number:  1253286-56-4
Price
710,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2,5-Dioxopyrrolidin-1-yl 14-oxo-18-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-13-azaoctadecanoate
  • CAS Number:1253286-56-4
  • Formula:C₂₃H₃₆N₄O₉S
  • MDL Number:MFCD11974977
  • Molecular Weight:544.618 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications