Biotin-PEG3-NHS ester
Bottle
Catalog # APOSBIPG1229-100MG
New Product
Supplier: Apollo Scientific
Biotin-PEG3-NHS ester
Catalog # APOSBIPG1229-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 1253286-56-4
Jump to:
Product Details & Documents
Synonyms: 2,5-Dioxopyrrolidin-1-yl 14-oxo-18-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-13-azaoctadecanoate
- CAS Number:1253286-56-4
- Formula:C₂₃H₃₆N₄O₉S
- MDL Number:MFCD11974977
- Molecular Weight:544.618 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



