D-(-)-Aspartic acid
Bottle
N° de catalogue APOSBIA0866-5KG
New Product
Fournisseur: Apollo Scientific
D-(-)-Aspartic acid
N° de catalogue APOSBIA0866-5KG
Fournisseur: Apollo Scientific
New Product
Numéro CAS: 1783-96-6
Jump to:
Product Details & Documents
Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.
Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- CAS Number:1783-96-6
- Formula:C₄H₇NO₄
- MDL Number:MFCD00063081
- Molecular Weight:133.1 g/mol
- Melting Pt:>300 °C
- Melting Pt:>300 °C
- Density:1.66 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:217-234-6
Specifications
Specifications
- Pack type:Bottle



