D-(-)-Aspartic acid
Bottle
Catalog # APOSBIA0866-500G
New Product
Supplier: Apollo Scientific
D-(-)-Aspartic acid
Catalog # APOSBIA0866-500G
Supplier: Apollo Scientific
New Product
CAS Number: 1783-96-6
Jump to:
Product Details & Documents
Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.
Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- CAS Number:1783-96-6
- Formula:C₄H₇NO₄
- MDL Number:MFCD00063081
- Molecular Weight:133.1 g/mol
- Melting Pt:>300 °C
- Melting Pt:>300 °C
- Density:1.66 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:217-234-6
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



