2-Ethylpiperidine 5g pack 1 * 5 g
Catalog # APOSOR911632-5G
Supplier: Apollo Scientific
New Product
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
VWR® Reagent Reservoirs
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Trifluoroacetic anhydride 99+%
Supplier: Thermo Fisher Scientific
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF