DINO-LITE EDGE+ DIGITAL MICROSCOPE 1.3MP 1 * 1 items
Catalog # IDCPAM4517MT-GRFBY
Fournisseur: Dino-Lite / IDCP B.V.
New Product
DINO-LITE EDGE+ DIGITAL MICROSCOPE 1.3MP 1 * 1 items
Catalog # IDCPAM4517MT-GRFBY
Fournisseur: Dino-Lite / IDCP B.V.
New Product
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Fournisseur: VWR Chemicals
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
VWR® Reagent Reservoirs
Fournisseur: VWR Collection
Pipette tips, Finntip™
Fournisseur: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Fournisseur: Merck
Ethanol absolute ≥99.5% USP, Ph. Eur.
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Fournisseur: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals