ContactUsLinkComponent
3 components: 20g/l each of Fe Pb Zn in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 250 mL
Catalog # CMPLRM081332L25
New Product
3 components: 20g/l each of Fe Pb Zn in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 250 mL
Catalog # CMPLRM081332L25