ContactUsLinkComponent
,,7 components: B 500mg/l Cu 500mg/l Fe 500mg/l Mn 500mg/l Mo 100mg/l Zn 500mg/l Ca 1000mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM118458L5
New Product
,,7 components: B 500mg/l Cu 500mg/l Fe 500mg/l Mn 500mg/l Mo 100mg/l Zn 500mg/l Ca 1000mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM118458L5