ContactUsLinkComponent
2D Tube Rack SAFE®, External thread, 4000µl, Non-Side Coded, Mulberry Capped, Automation Friendly Rack, Beta-Irradiated. (50x24) 1 * 1 items
Catalog # ELKAAZ3C1-XL4-MU-A
Supplier:  ELKAY
New Product
2D Tube Rack SAFE®, External thread, 4000µl, Non-Side Coded, Mulberry Capped, Automation Friendly Rack, Beta-Irradiated. (50x24) 1 * 1 items
Catalog # ELKAAZ3C1-XL4-MU-A
Supplier:  ELKAY