13 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM117926L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
13 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM117926L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR Collection
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Water standards, CRM, Karl Fischer titration, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals