ContactUsLinkComponent
DINO-LITE, DIGITAL MICROSCOPE, 1.3MP, 20x-50x & 200x MAGNIFICATION, MEASUREMENT FUNCTIONALITY, WIRELESS READY (MODULE SOLD SEPARATELY) 1 * 1 items
Catalog # IDCPAF3113T
Fournisseur:  Dino-Lite / IDCP B.V.
New Product
DINO-LITE, DIGITAL MICROSCOPE, 1.3MP, 20x-50x & 200x MAGNIFICATION, MEASUREMENT FUNCTIONALITY, WIRELESS READY (MODULE SOLD SEPARATELY) 1 * 1 items
Catalog # IDCPAF3113T
Fournisseur:  Dino-Lite / IDCP B.V.