ContactUsLinkComponent
Comfort MTP racks for chest freezers for 13 SBS format plates, 55mm high, stainless steel, dimensions (HxDxW) 739x135x92mm 1 * 1 items
Catalog # TENATE35276
Supplier:  TENAK
New Product
Comfort MTP racks for chest freezers for 13 SBS format plates, 55mm high, stainless steel, dimensions (HxDxW) 739x135x92mm 1 * 1 items
Catalog # TENATE35276
Supplier:  TENAK