Comfort rack for upright freezers for (HxD) 4x3=12 cryoboxes, 75mm high, stainless steel, sliding shelves, dimensions (HxDxW) 319x42 4x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24266B10
Supplier: TENAK