ContactUsLinkComponent
Eco AluRack for chest freezers for 12 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 437x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11462B10
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 12 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 437x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11462B10
Supplier:  TENAK