Eco AluRack for upright freezers for (HxD) 3x3=9 cryoboxes, 130mm high, aluminium, dimensions (HxDxW) 401x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11346B10
Supplier: TENAK