ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 3x4=12 cryoboxes, 50mm high, aluminium, dimensions (HxDxW) 168x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11230B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 3x4=12 cryoboxes, 50mm high, aluminium, dimensions (HxDxW) 168x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11230B10
Supplier:  TENAK