ContactUsLinkComponent
Product developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplan tation genetic testing 1 * 1 KIT
Catalog # BIOO4341-IL48A
Supplier:  BIOO SCIENTIFIC
undefined
Product developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplan tation genetic testing 1 * 1 KIT
Catalog # BIOO4341-IL48A
Supplier:  BIOO SCIENTIFIC
Frequently Bought Together