Order Entry
Belgium
Orders LinkContactUsLinkComponent
NEXTFLEX™ Universal Blockers

NEXTFLEX™ Universal Blockers

NEXTFLEX™ universal blockers, 8
Catalog # BIOONOVA-5143231
Marketsource
Supplier:   Revvity
NEXTFLEX™ Universal Blockers
Catalog # BIOONOVA-5143231
Supplier:  Revvity
Marketsource
CAS Number:  
NEXTFLEX™ Universal Blockers
NEXTFLEX™ Universal Blockers

NEXTFLEX™ universal blockers, 8

CAS Number:  
Price
103,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

The NEXTFLEX™ universal blockers block non-specific hybridisation, enhancing specificity and reducing sequencing costs. They are compatible with all ligation and tagmentation workflows for Illumina®.

  • Effective in blocking single-index or dual-index libraries Compatible with all NEXTFLEX™ barcodes and all other ligation-based and tagmentation-based workflows for Illumina® sequencing platforms Simplifies and reduces the cost of target capture without sacrificing performance 96 barcode blockers available

During target enrichment, adapters can interact with the complementary adapter sequence strand of another library molecule. The NEXTFLEX™ universal blockers are designed to prevent this concatemerisation of library molecules. By blocking this complementary adapter sequence interaction, the number of on-target reads improves dramatically and the depth of enrichment increases. The NEXTFLEX™ universal blockers are designed for use with indexed adapters for Illumina sequencing platforms. They are effective in blocking single-indexed or dual-indexed libraries, without being affected by index length or the presence of a unique molecular index (UMI). The NEXTFLEX Universal Blockers are compatible with ligation-based workflows, such as NEXTFLEX™ or Illumina trusequence library prep kits. They are also compatible with tagmentation-based workflows, such as Illumina Nextera library prep kits.

Certificeringen: Manufactured in ISO 9001 facility.

Let op: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ universal blockers
  • No. of reactions:
    8

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.